siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 5 | 703 | |||
| NR_028410.1 | 4 | 672 | ||||
| NR_028411.1 | 4 | 636 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00207| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 5 | 703 | |||
| NR_028410.1 | 4 | 672 | ||||
| NR_028411.1 | 4 | 636 | ||||