siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 5 | 469 | |||
| NR_028410.1 | 4 | 438 | ||||
| NR_028411.1 | 4 | 402 | ||||
| GenBank mRNA | BC144506.1 | - | 4 | 426 | ||
| BC144508.1 | - | 5 | 457 | |||
| BU585030.1 | - | 4 | 421 | |||

LINC00207
Genomic Map