siRNA Details
Gene Symbol: ANKRD26P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027248.3 | 14 | 2029 | |||
GenBank mRNA | BC036210.1 | - | 14 | 2029 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ANKRD26P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027248.3 | 14 | 2029 | |||
GenBank mRNA | BC036210.1 | - | 14 | 2029 |