siRNA Details
Gene Symbol: LOC642929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027472.1 | 5 | 949 | |||
GenBank mRNA | DQ786189.1 | - | 5 | 948 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC642929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027472.1 | 5 | 949 | |||
GenBank mRNA | DQ786189.1 | - | 5 | 948 |