siRNA Details
| Gene Symbol: LOC642929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027472.1 | 3 | 512 | |||
| GenBank mRNA | DQ786189.1 | - | 3 | 511 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX096823.1 | - | 3 | 537 | Chr. 9 - 66162117 - 66164929 [-] on Build GRCh38 | |

LOC642929
Genomic Map