siRNA Details
Gene Symbol: LOC642929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027472.1 | 2, 3 | 480 | |||
GenBank mRNA | DQ786189.1 | - | 2, 3 | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX096823.1 | - | 2, 3 | 505 | Chr. 9 - 66162117 - 66164929 [-] on Build GRCh38 |
