siRNA Details
| Gene Symbol: LINC00442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026852.1 | 1 | 325 | |||
| GenBank mRNA | AK301801.1 | - | 1 | 325 | ||
| BC047425.1 | - | 1 | 325 | |||
| BC067875.1 | - | 1 | 325 | |||
| BC094789.1 | - | 1 | 325 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456737.1 | - | 1 | 325 | Chr. 13 - 19008259 - 19012559 [+] on Build GRCh38 | |

LINC00442
Genomic Map