siRNA Details
| Gene Symbol: FLVCR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027285.1 | 2 | 858 | |||
| NR_027286.1 | 2 | 604 | ||||
| GenBank mRNA | AY030238.1 | - | 2 | 797 | ||
| AY030239.1 | - | 2 | 543 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF686299.1 | - | 2 | 641 | Chr. 1 - 212856632 - 212857911 [-] on Build GRCh38 | |

FLVCR1-AS1
Genomic Map