siRNA Details
Gene Symbol: LOH12CR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024061.1 | 2 | 1109 | |||
GenBank mRNA | AY037867.1 | - | 2 | 1109 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB049004.1 | - | 1 | 171 | Chr. 12 - 12355517 - 12355999 [-] on Build GRCh38 |
