siRNA Details
| Gene Symbol: SMIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028439.1 | 1 | 520 | |||
| XM_017024945.1 | XP_016880434.1 | 2 | 270 | |||
| GenBank mRNA | AL832643.2 | - | 1 | 520 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM5| Gene Symbol: SMIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028439.1 | 1 | 520 | |||
| XM_017024945.1 | XP_016880434.1 | 2 | 270 | |||
| GenBank mRNA | AL832643.2 | - | 1 | 520 | ||