siRNA Details
| Gene Symbol: CDRT15P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003261.1 | 1 | 85 | |||
| GenBank mRNA | BX118788.1 | - | 1 | 86 | ||
| Gene Symbol: CDRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC071823.1 | - | 1 | 101 | ||

CDRT15P1
Genomic Map