siRNA Details
| Gene Symbol: H1FX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026991.1 | 1 | 988 | |||
| GenBank mRNA | AL353963.1 | - | 1 | 280 | ||
| BM709882.1 | - | 1 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H1FX-AS1| Gene Symbol: H1FX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026991.1 | 1 | 988 | |||
| GenBank mRNA | AL353963.1 | - | 1 | 280 | ||
| BM709882.1 | - | 1 | 214 | |||