siRNA Details
| Gene Symbol: PCED1B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026544.1 | 3 | 1783 | |||
| GenBank mRNA | AK123977.1 | - | 3 | 1775 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCED1B-AS1| Gene Symbol: PCED1B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026544.1 | 3 | 1783 | |||
| GenBank mRNA | AK123977.1 | - | 3 | 1775 | ||