siRNA Details
| Gene Symbol: FBXO22-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003136.1 | 1 | 136 | |||
| GenBank mRNA | AA843761.1 | - | 1 | 140 | ||
| AA931120.1 | - | 1 | 141 | |||
| AI868357.1 | - | 1 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO22-AS1| Gene Symbol: FBXO22-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003136.1 | 1 | 136 | |||
| GenBank mRNA | AA843761.1 | - | 1 | 140 | ||
| AA931120.1 | - | 1 | 141 | |||
| AI868357.1 | - | 1 | 140 | |||