siRNA Details
| Gene Symbol: PITPNA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028514.1 | 1, 2 | 177 | |||
| GenBank mRNA | AA341969.1 | - | 1, 2 | 164 | ||
| DB053306.1 | - | 1, 2 | 178 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PITPNA-AS1| Gene Symbol: PITPNA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028514.1 | 1, 2 | 177 | |||
| GenBank mRNA | AA341969.1 | - | 1, 2 | 164 | ||
| DB053306.1 | - | 1, 2 | 178 | |||