siRNA Details
| Gene Symbol: LOC100126784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015384.2 | 1 | 3661 | |||
| GenBank mRNA | BC047636.1 | - | 1 | 1491 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM714322.1 | - | 1 | 637 | Chr. 11 - 19710945 - 19711648 [-] on Build GRCh38 | |

LOC100126784
Genomic Map