siRNA Details
| Gene Symbol: LOC100126784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015384.2 | 1 | 2919 | |||
| GenBank mRNA | BC037245.1 | - | 1 | 866 | ||
| BC047636.1 | - | 1 | 749 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100126784| Gene Symbol: LOC100126784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015384.2 | 1 | 2919 | |||
| GenBank mRNA | BC037245.1 | - | 1 | 866 | ||
| BC047636.1 | - | 1 | 749 | |||