siRNA Details
| Gene Symbol: LOC100126784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015384.2 | 1 | 301 | |||
| GenBank mRNA | BC029949.1 | - | 1 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100126784| Gene Symbol: LOC100126784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015384.2 | 1 | 301 | |||
| GenBank mRNA | BC029949.1 | - | 1 | 192 | ||