siRNA Details
| Gene Symbol: CYMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003599.2 | 8 | 949 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYMP| Gene Symbol: CYMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003599.2 | 8 | 949 | |||