siRNA Details
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023380.1 | 14, 15 | 4025 | |||
GenBank mRNA | AK295707.1 | - | 2, 3 | 338 | ||
DA321951.1 | - | 2, 3 | 338 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023380.1 | 14, 15 | 4025 | |||
GenBank mRNA | AK295707.1 | - | 2, 3 | 338 | ||
DA321951.1 | - | 2, 3 | 338 |