siRNA Details
| Gene Symbol: ZFHX4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024360.1 | 4 | 1540 | |||
| GenBank mRNA | BC037827.1 | - | 4 | 1548 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFHX4-AS1| Gene Symbol: ZFHX4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024360.1 | 4 | 1540 | |||
| GenBank mRNA | BC037827.1 | - | 4 | 1548 | ||