siRNA Details
Gene Symbol: PP12613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024365.1 | 1 | 1902 | |||
GenBank mRNA | AF318325.1 | - | 1 | 1903 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PP12613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024365.1 | 1 | 1902 | |||
GenBank mRNA | AF318325.1 | - | 1 | 1903 |