siRNA Details
| Gene Symbol: FLJ16779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024389.1 | 1 | 2741 | |||
| GenBank mRNA | AK131543.1 | - | 1 | 2741 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ16779| Gene Symbol: FLJ16779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024389.1 | 1 | 2741 | |||
| GenBank mRNA | AK131543.1 | - | 1 | 2741 | ||