siRNA Details
| Gene Symbol: MFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242336.1 | NP_001229265.1 | 1 | 120 | ||
| GenBank mRNA | AL037715.1 | - | 1 | 89 | ||
| BC026244.1 | - | 1 | 80 | 4697443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFAP3| Gene Symbol: MFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242336.1 | NP_001229265.1 | 1 | 120 | ||
| GenBank mRNA | AL037715.1 | - | 1 | 89 | ||
| BC026244.1 | - | 1 | 80 | 4697443 | ||