siRNA Details
| Gene Symbol: LOC389332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024418.1 | 1 | 1227 | |||
| GenBank mRNA | BC031291.1 | - | 1 | 1111 | ||
| BC048329.1 | - | 1 | 1198 | |||
| BC062612.1 | - | 1 | 1185 | |||
| BC073887.1 | - | 1 | 1185 | |||
| BC092501.1 | - | 1 | 1111 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI791392.1 | - | 1 | 55 | Chr. 5 - 136191600 - 136191991 [-] on Build GRCh38 | |

LOC389332
Genomic Map