siRNA Details
| Gene Symbol: LOC389332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024418.1 | 1 | 1438 | |||
| GenBank mRNA | BC031291.1 | - | 1 | 1322 | ||
| BC048329.1 | - | 1 | 1409 | |||
| BC062612.1 | - | 1 | 1396 | |||
| BC073887.1 | - | 1 | 1396 | |||
| BC092501.1 | - | 1 | 1322 | |||

LOC389332
Genomic Map