siRNA Details
| Gene Symbol: FLJ23867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026900.1 | 1 | 1290 | |||
| GenBank mRNA | AK074447.1 | - | 1 | 1310 | ||
| AL832927.1 | - | 1 | 1290 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ23867| Gene Symbol: FLJ23867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026900.1 | 1 | 1290 | |||
| GenBank mRNA | AK074447.1 | - | 1 | 1310 | ||
| AL832927.1 | - | 1 | 1290 | |||