siRNA Details
| Gene Symbol: MOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003090.2 | 6 | 2052 | |||
| NR_103504.1 | 7 | 2394 | ||||
| NR_103505.1 | 7 | 2432 | ||||
| GenBank mRNA | BC022471.1 | - | 6 | 1947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOBP| Gene Symbol: MOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003090.2 | 6 | 2052 | |||
| NR_103504.1 | 7 | 2394 | ||||
| NR_103505.1 | 7 | 2432 | ||||
| GenBank mRNA | BC022471.1 | - | 6 | 1947 | ||