siRNA Details
| Gene Symbol: LOC143666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026967.1 | 1 | 1520 | |||
| GenBank mRNA | AK057794.1 | - | 2 | 1103 | ||
| AK123970.1 | - | 1 | 1519 | |||
| BC018884.2 | - | 1 | 497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC143666| Gene Symbol: LOC143666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026967.1 | 1 | 1520 | |||
| GenBank mRNA | AK057794.1 | - | 2 | 1103 | ||
| AK123970.1 | - | 1 | 1519 | |||
| BC018884.2 | - | 1 | 497 | |||