siRNA Details
| Gene Symbol: LOC143666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026967.1 | 1 | 1657 | |||
| GenBank mRNA | AK057794.1 | - | 2 | 1240 | ||
| AK123970.1 | - | 1 | 1656 | |||
| BC018884.2 | - | 1 | 634 | |||
| Gene Symbol: LOC100288175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC033949.2 | - | 4 | 454 | ||

LOC143666
Genomic Map