siRNA Details
| Gene Symbol: FAM183BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028347.1 | 1 | 255 | |||
| GenBank mRNA | AK124132.1 | - | 1 | 255 | ||
| BC045803.1 | - | 1 | 203 | |||
| BG772121.1 | - | 1 | 208 | |||
| DB047796.1 | - | 1 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM183BP| Gene Symbol: FAM183BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028347.1 | 1 | 255 | |||
| GenBank mRNA | AK124132.1 | - | 1 | 255 | ||
| BC045803.1 | - | 1 | 203 | |||
| BG772121.1 | - | 1 | 208 | |||
| DB047796.1 | - | 1 | 255 | |||