siRNA Details
| Gene Symbol: PRO0611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI064823.1 | - | 1 | 402 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF111851.1 | - | 402 | Chr. - - [-] on Build | ||

PRO0611
Genomic Map