siRNA Details
| Gene Symbol: LOC643406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029405.1 | 1 | 28 | |||
| GenBank mRNA | AK125948.1 | - | 1 | 18 | ||
| BC031676.1 | - | 1 | 17 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643406| Gene Symbol: LOC643406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029405.1 | 1 | 28 | |||
| GenBank mRNA | AK125948.1 | - | 1 | 18 | ||
| BC031676.1 | - | 1 | 17 | |||