siRNA Details
| Gene Symbol: FOXN3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024620.1 | 1 | 624 | |||
| GenBank mRNA | AF118074.1 | - | 1 | 624 | ||
| BC069658.1 | - | 1 | 569 | |||
| BC069757.1 | - | 1 | 573 | |||
| BC069775.1 | - | 1 | 573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXN3-AS2| Gene Symbol: FOXN3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024620.1 | 1 | 624 | |||
| GenBank mRNA | AF118074.1 | - | 1 | 624 | ||
| BC069658.1 | - | 1 | 569 | |||
| BC069757.1 | - | 1 | 573 | |||
| BC069775.1 | - | 1 | 573 | |||