siRNA Details
| Gene Symbol: MIR1250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031652.1 | 1 | 81 | |||
| Gene Symbol: AATK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525509.2 | XP_011523811.1 | 1 | 1126 | ||

MIR1250
Genomic Map