siRNA Details
Gene Symbol: FLJ37201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026835.1 | 2 | 2002 | |||
GenBank mRNA | AK094520.1 | - | 1 | 1157 | ||
BC043217.2 | - | 2 | 2001 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ37201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026835.1 | 2 | 2002 | |||
GenBank mRNA | AK094520.1 | - | 1 | 1157 | ||
BC043217.2 | - | 2 | 2001 |