siRNA Details
| Gene Symbol: RNU6ATAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023344.1 | 1 | 81 | |||
| GenBank mRNA | U62823.1 | - | 1 | 81 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNU6ATAC| Gene Symbol: RNU6ATAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023344.1 | 1 | 81 | |||
| GenBank mRNA | U62823.1 | - | 1 | 81 | ||