siRNA Details
| Gene Symbol: RNU6ATAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023344.1 | 1 | 22 | |||
| GenBank mRNA | U62823.1 | - | 1 | 22 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK024534.1 | - | 1 | 527 | Chr. 17 - 31562310 - 31564399 [-] on Build GRCh38 | |

RNU6ATAC
Genomic Map