siRNA Details
| Gene Symbol: ASAP1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002765.2 | 1 | 357 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF150205.1 | - | 1 | 355 | Chr. 8 - 130295355 - 130296523 [-] on Build GRCh38 | |

ASAP1-IT1
Genomic Map