siRNA Details
Gene Symbol: ASAP1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002765.2 | 1 | 692 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF150205.1 | - | 1 | 688 | Chr. 8 - 130295355 - 130296523 [-] on Build GRCh38 |
