siRNA Details
| Gene Symbol: LINC00652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026883.1 | 4 | 1419 | |||
| NR_026884.1 | 3 | 1242 | ||||
| GenBank mRNA | AK094606.1 | - | 3 | 1240 | ||
| BC029555.1 | - | 3 | 1252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00652| Gene Symbol: LINC00652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026883.1 | 4 | 1419 | |||
| NR_026884.1 | 3 | 1242 | ||||
| GenBank mRNA | AK094606.1 | - | 3 | 1240 | ||
| BC029555.1 | - | 3 | 1252 | |||