siRNA Details
| Gene Symbol: LINC00094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015427.2 | 3 | 1440 | |||
| GenBank mRNA | AK092667.1 | - | 2 | 1298 | ||
| BC094875.1 | - | 1 | 788 | |||
| BC108326.1 | - | 1 | 1096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00094| Gene Symbol: LINC00094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015427.2 | 3 | 1440 | |||
| GenBank mRNA | AK092667.1 | - | 2 | 1298 | ||
| BC094875.1 | - | 1 | 788 | |||
| BC108326.1 | - | 1 | 1096 | |||