siRNA Details
| Gene Symbol: ZNF788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027049.1 | 3 | 610 | |||
| GenBank mRNA | AK128700.1 | - | 3 | 610 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN263918.1 | - | 4 | 567 | Chr. 19 - 12092385 - 12111660 [+] on Build GRCh38 | |

ZNF788
Genomic Map