siRNA Details
Gene Symbol: ESRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027122.1 | 4 | 2921 | |||
GenBank mRNA | DQ445779.2 | - | 4 | 2921 | ||
KM363598.1 | - | 4 | 2617 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ESRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027122.1 | 4 | 2921 | |||
GenBank mRNA | DQ445779.2 | - | 4 | 2921 | ||
KM363598.1 | - | 4 | 2617 |