siRNA Details
| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015429.1 | 8 | 1652 | |||
| NR_024429.1 | 8 | 1748 | ||||
| NR_024431.2 | 10 | 1944 | ||||
| GenBank mRNA | AK097923.1 | - | 4 | 2363 | ||
| AK098249.1 | - | 10 | 2389 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456353.5 | - | 10 | 2391 | Chr. 10 - 78943328 - 79067895 [-] on Build GRCh38 | |

ZMIZ1-AS1
Genomic Map