siRNA Details
| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015429.1 | 8 | 1911 | |||
| NR_024429.1 | 8 | 2007 | ||||
| NR_024431.2 | 10 | 2203 | ||||
| GenBank mRNA | AK097923.1 | - | 4 | 2622 | ||
| AK098249.1 | - | 10 | 2648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZMIZ1-AS1| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015429.1 | 8 | 1911 | |||
| NR_024429.1 | 8 | 2007 | ||||
| NR_024431.2 | 10 | 2203 | ||||
| GenBank mRNA | AK097923.1 | - | 4 | 2622 | ||
| AK098249.1 | - | 10 | 2648 | |||