siRNA Details
Gene Symbol: LINC00691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026834.1 | 1 | 139 | |||
GenBank mRNA | BC033355.1 | - | 1 | 130 | ||
BC045572.1 | - | 1 | 115 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026834.1 | 1 | 139 | |||
GenBank mRNA | BC033355.1 | - | 1 | 130 | ||
BC045572.1 | - | 1 | 115 |