siRNA Details
| Gene Symbol: SMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029426.1 | 10 | 1800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMA4| Gene Symbol: SMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029426.1 | 10 | 1800 | |||