siRNA Details
| Gene Symbol: KRT8P41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027713.1 | 1 | 1540 | |||
| GenBank mRNA | AK127993.1 | - | 1 | 1540 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT8P41| Gene Symbol: KRT8P41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027713.1 | 1 | 1540 | |||
| GenBank mRNA | AK127993.1 | - | 1 | 1540 | ||