siRNA Details
| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028796.1 | XP_016884285.1 | 6 | 5714 | ||
| GenBank mRNA | AK126430.1 | - | 5 | 2746 | ||
| BC048207.1 | - | 4 | 1401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENSG00000188511
C22orf34| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028796.1 | XP_016884285.1 | 6 | 5714 | ||
| GenBank mRNA | AK126430.1 | - | 5 | 2746 | ||
| BC048207.1 | - | 4 | 1401 | |||